4-Chloro-2-fluoro-N-methylbenzamide 98%

Lieferant: Apollo Scientific

PC910133-1G
APOSPC910133-1GEA 71.2 EUR
APOSPC910133-1G
4-Chloro-2-fluoro-N-methylbenzamide 98%
4-Chloro-2-fluoro-N-methylbenzamide
Formel: C₈H₇ClFNO
Molecular Weight: 187,599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19596644
CAS-Nummer: 1343038-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR