2-Ethoxy-3-fluorophenol 95%

Lieferant: Apollo Scientific

PC910221-1G
APOSPC910221-1GEA 632 EUR
APOSPC910221-1G
2-Ethoxy-3-fluorophenol 95%
2-Ethoxy-3-fluorophenol
Formel: C₈H₉FO₂
Molecular Weight: 156,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23111044
CAS-Nummer: 909302-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR