2-Ethoxy-6-fluoro-phenol 95%

Lieferant: Apollo Scientific

Synonyme: 2-Ethoxy-6-fluorophenol

PC910223-1G
APOSPC910223-1GEA 632 EUR
APOSPC910223-1G
2-Ethoxy-6-fluoro-phenol 95%
2-Ethoxy-6-fluoro-phenol
Formel: C₈H₉FO₂
Molecular Weight: 156,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16878639
CAS-Nummer: 868761-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR