5-Fluororesorcinol 95%
Lieferant: Apollo Scientific
Synonyme:
5-Fluorobenzene-1,3-diol
PC910226-1G
APOSPC910226-1GEA
133
EUR
APOSPC910226-1G
5-Fluororesorcinol 95%
5-Fluororesorcinol
Formel:
C₆H₅FO₂ Molecular Weight: 128,10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08457182 CAS-Nummer: 75996-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...