4-Fluoro-2-(hydroxymethyl)phenol 95%

Lieferant: Apollo Scientific

PC910230-1G
APOSPC910230-1GEA 179 EUR
APOSPC910230-1G
4-Fluoro-2-(hydroxymethyl)phenol 95%
4-Fluoro-2-(hydroxymethyl)phenol
Formel: C₇H₇FO₂
Molecular Weight: 142,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412237
CAS-Nummer: 2357-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR