2-Fluoro-5-hydroxybenzyl alcohol 95%

Lieferant: Apollo Scientific

PC910232-1G
APOSPC910232-1GEA 0 EUR
APOSPC910232-1G
2-Fluoro-5-hydroxybenzyl alcohol 95%
2-Fluoro-5-hydroxybenzyl alcohol
Formel: C₇H₇FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12923295
CAS-Nummer: 438049-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR