6-Bromo-2-fluoropyridin-3-ol 95%

Lieferant: Apollo Scientific

PC910235-1G
APOSPC910235-1GEA 0 EUR
APOSPC910235-1G
6-Bromo-2-fluoropyridin-3-ol 95%
6-Bromo-2-fluoropyridin-3-ol
Formel: C₅H₃BrFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09999156
CAS-Nummer: 850142-72-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR