5-Ethoxy-2-fluorophenol 95%

Lieferant: Apollo Scientific

PC910236-1G
APOSPC910236-1GEA 632 EUR
APOSPC910236-1G
5-Ethoxy-2-fluorophenol 95%
5-Ethoxy-2-fluorophenol
Formel: C₈H₉FO₂
Molecular Weight: 156,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18974322
CAS-Nummer: 577793-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR