3,5-Difluoro-2-methoxyphenol 96%

Lieferant: Apollo Scientific

Synonyme: 2,4-Difluoro-6-hydroxyanisole

PC910240-1G
APOSPC910240-1GEA 364 EUR
APOSPC910240-1G
3,5-Difluoro-2-methoxyphenol 96%
3,5-Difluoro-2-methoxyphenol
Formel: C₇H₆F₂O₂
Molecular Weight: 160,118 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12404933
CAS-Nummer: 152434-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR