2,5-Difluoro-4-methoxyphenol 95%
Lieferant: Apollo Scientific
PC910241-1G
APOSPC910241-1GEA
0
EUR
APOSPC910241-1G
2,5-Difluoro-4-methoxyphenol 95%
2,5-Difluoro-4-methoxyphenol
Formel:
C₇H₆F₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06246893 CAS-Nummer: 1024068-86-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...