6-Bromo-2,3-difluorophenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC910246-1G
PC910246-5G
APOSPC910246-1GEA
255
EUR
APOSPC910246-1G
APOSPC910246-5G
6-Bromo-2,3-difluorophenol 97%
6-Bromo-2,3-difluorophenol
Formel:
C₆H₃BrF₂O Lagertemperatur: Kühlschrank |
MDL:
MFCD11656842 CAS-Nummer: 186590-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...