2-Fluoro-3-(propan-2-yloxy)phenol 95%

Lieferant: Apollo Scientific

PC910247-1G
APOSPC910247-1GEA 632 EUR
APOSPC910247-1G
2-Fluoro-3-(propan-2-yloxy)phenol 95%
2-Fluoro-3-(propan-2-yloxy)phenol
Formel: C₉H₁₁FO₂
Molecular Weight: 170,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16997428
CAS-Nummer: 1243452-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR