3-Fluoro-4-(propan-2-yloxy)phenol 96%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-4-isopropoxyphenol

PC910248-1G
APOSPC910248-1GEA 364 EUR
APOSPC910248-1G
3-Fluoro-4-(propan-2-yloxy)phenol 96%
3-Fluoro-4-(propan-2-yloxy)phenol
Formel: C₉H₁₁FO₂
Molecular Weight: 170,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16997390
CAS-Nummer: 1243280-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR