3-Chloro-5-fluoro-2-methoxyphenol 95%

Lieferant: Apollo Scientific

PC910250-1G
APOSPC910250-1GEA 632 EUR
APOSPC910250-1G
3-Chloro-5-fluoro-2-methoxyphenol 95%
3-Chloro-5-fluoro-2-methoxyphenol
Formel: C₇H₆ClFO₂
Molecular Weight: 176,573 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22415068
CAS-Nummer: 1783971-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR