3-Chloro-4-ethoxy-2-fluorophenol 95%

Lieferant: Apollo Scientific

PC910255-1G
APOSPC910255-1GEA 632 EUR
APOSPC910255-1G
3-Chloro-4-ethoxy-2-fluorophenol 95%
3-Chloro-4-ethoxy-2-fluorophenol
Formel: C₈H₈ClFO₂
Molecular Weight: 190,599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24728516
CAS-Nummer: 909122-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR