1-Hydroxy-4-propyloxy-2,3-difluorobenzene 95%

Lieferant: Apollo Scientific

PC910256-1G
APOSPC910256-1GEA 632 EUR
APOSPC910256-1G
1-Hydroxy-4-propyloxy-2,3-difluorobenzene 95%
1-Hydroxy-4-propyloxy-2,3-difluorobenzene
Formel: C₉H₁₀F₂O₂
Molecular Weight: 188,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368024
CAS-Nummer: 887582-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR