4-Fluoro-2-methoxy-5-methylphenol 98%
Lieferant: Apollo Scientific
PC910257-1G
APOSPC910257-1GEA
364
EUR
APOSPC910257-1G
4-Fluoro-2-methoxy-5-methylphenol 98%
4-Fluoro-2-methoxy-5-methylphenol
Formel:
C₈H₉FO₂ Molecular Weight: 156,154 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23110601 CAS-Nummer: 1395035-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...