3,4-Difluoro-5-methoxyphenol 95%
Lieferant: Apollo Scientific
PC910259-1G
APOSPC910259-1GEA
632
EUR
APOSPC910259-1G
3,4-Difluoro-5-methoxyphenol 95%
3,4-Difluoro-5-methoxyphenol
Formel:
C₇H₆F₂O₂ Molecular Weight: 160,118 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22414498 CAS-Nummer: 1394915-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...