3-Chloro-2,4-difluorophenol 95%

Lieferant: Apollo Scientific

PC910261-1G
APOSPC910261-1GEA 632 EUR
APOSPC910261-1G
3-Chloro-2,4-difluorophenol 95%
3-Chloro-2,4-difluorophenol
Formel: C₆H₃ClF₂O
Molecular Weight: 164,537 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415629
CAS-Nummer: 2268-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR