4-Chloro-2-fluororesorcinol 95%
Lieferant: Apollo Scientific
PC910262-1G
APOSPC910262-1GEA
632
EUR
APOSPC910262-1G
4-Chloro-2-fluororesorcinol 95%
4-Chloro-2-fluororesorcinol
Formel:
C₆H₄ClFO₂ Molecular Weight: 162,546 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23713137 CAS-Nummer: 1070868-27-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...