2-Fluoro-5-(propan-2-yloxy)phenol 96%

Lieferant: Apollo Scientific

PC910263-1G
APOSPC910263-1GEA 364 EUR
APOSPC910263-1G
2-Fluoro-5-(propan-2-yloxy)phenol 96%
2-Fluoro-5-(propan-2-yloxy)phenol
Formel: C₉H₁₁FO₂
Molecular Weight: 170,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16997357
CAS-Nummer: 1243289-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR