4-Ethoxy-2,5-difluorophenol 95%

Lieferant: Apollo Scientific

PC910266-1G
APOSPC910266-1GEA 632 EUR
APOSPC910266-1G
4-Ethoxy-2,5-difluorophenol 95%
4-Ethoxy-2,5-difluorophenol
Formel: C₈H₈F₂O₂
Molecular Weight: 174,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24728303
CAS-Nummer: 612092-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR