2,4-Difluoro-3-methoxy-5-methylphenol 95%

Lieferant: Apollo Scientific

PC910268-1G
APOSPC910268-1GEA 632 EUR
APOSPC910268-1G
2,4-Difluoro-3-methoxy-5-methylphenol 95%
2,4-Difluoro-3-methoxy-5-methylphenol
Formel: C₈H₈F₂O₂
Molecular Weight: 174,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24730019
CAS-Nummer: 1422360-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR