3-Fluoro-5-(propan-2-yloxy)phenol 95%

Lieferant: Apollo Scientific

PC910269-1G
APOSPC910269-1GEA 632 EUR
APOSPC910269-1G
3-Fluoro-5-(propan-2-yloxy)phenol 95%
3-Fluoro-5-(propan-2-yloxy)phenol
Formel: C₉H₁₁FO₂
Molecular Weight: 170,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16997310
CAS-Nummer: 1243373-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR