3,5-Dichloro-4-fluorophenol 96%

Lieferant: Apollo Scientific

PC910271-1G
APOSPC910271-1GEA 411 EUR
APOSPC910271-1G
3,5-Dichloro-4-fluorophenol 96%
3,5-Dichloro-4-fluorophenol
Formel: C₆H₃Cl₂FO
Molecular Weight: 180,992 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19690558
CAS-Nummer: 2995-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR