3-Hydroxy-2-methoxy-5-(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific

PC910276-1G
APOSPC910276-1GEA 632 EUR
APOSPC910276-1G
3-Hydroxy-2-methoxy-5-(trifluoromethyl)pyridine 95%
3-Hydroxy-2-methoxy-5-(trifluoromethyl)pyridine
Formel: C₇H₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16611384
CAS-Nummer: 1227580-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR