2-Fluoro-4-hydroxy-N-methylbenzamide 95%

Lieferant: Apollo Scientific

PC910278-1G
APOSPC910278-1GEA 632 EUR
APOSPC910278-1G
2-Fluoro-4-hydroxy-N-methylbenzamide 95%
2-Fluoro-4-hydroxy-N-methylbenzamide
Formel: C₈H₈FNO₂
Molecular Weight: 169,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16998328
CAS-Nummer: 1243392-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR