2-Methoxy-3-(trifluoromethyl)phenol 95%

Lieferant: Apollo Scientific

PC910281-1G
APOSPC910281-1GEA 632 EUR
APOSPC910281-1G
2-Methoxy-3-(trifluoromethyl)phenol 95%
2-Methoxy-3-(trifluoromethyl)phenol
Formel: C₈H₇F₃O₂
Molecular Weight: 192,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185692
CAS-Nummer: 1214334-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR