2-Chloro-6-(trifluoromethyl)pyridin-3-ol 95%

Lieferant: Apollo Scientific

PC910286-1G
APOSPC910286-1GEA 172 EUR
APOSPC910286-1G
2-Chloro-6-(trifluoromethyl)pyridin-3-ol 95%
2-Chloro-6-(trifluoromethyl)pyridin-3-ol
Formel: C₆H₃ClF₃NO
Molecular Weight: 197,542 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16606449
CAS-Nummer: 731002-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR