2,5-Dihydroxybenzotrifluoride 95%
Lieferant: Apollo Scientific
PC910294-1G
APOSPC910294-1GEA
632
EUR
APOSPC910294-1G
2,5-Dihydroxybenzotrifluoride 95%
2,5-Dihydroxybenzotrifluoride
Formel:
C₇H₅F₃O₂ Molecular Weight: 178.109 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18395729 CAS-Nummer: 577-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...