5-(Trifluoromethyl)benzene-1,3-diol 95%
Lieferant: Apollo Scientific
PC910295-1G
APOSPC910295-1GEA
835
EUR
APOSPC910295-1G
5-(Trifluoromethyl)benzene-1,3-diol 95%
5-(Trifluoromethyl)benzene-1,3-diol
Formel:
C₇H₅F₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061289 CAS-Nummer: 454-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...