3-Methoxy-4-(trifluoromethyl)phenol 98%

Lieferant: Apollo Scientific

PC910297-1G
APOSPC910297-1GEA 364 EUR
APOSPC910297-1G
3-Methoxy-4-(trifluoromethyl)phenol 98%
3-Methoxy-4-(trifluoromethyl)phenol
Formel: C₈H₇F₃O₂
Molecular Weight: 192,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185696
CAS-Nummer: 106877-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR