4-(Trifluoromethoxy)benzene-1,3-diol 95%

Lieferant: Apollo Scientific

PC910300-1G
APOSPC910300-1GEA 632 EUR
APOSPC910300-1G
4-(Trifluoromethoxy)benzene-1,3-diol 95%
4-(Trifluoromethoxy)benzene-1,3-diol
Formel: C₇H₅F₃O₃
Molecular Weight: 194,108 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23111252
CAS-Nummer: 1395038-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR