1-(4-Fluorophenyl)-1H-pyrazol-4-ol 95%

Lieferant: Apollo Scientific

PC910301-1G
APOSPC910301-1GEA 364 EUR
APOSPC910301-1G
1-(4-Fluorophenyl)-1H-pyrazol-4-ol 95%
1-(4-Fluorophenyl)-1H-pyrazol-4-ol
Formel: C₉H₇FN₂O
Molecular Weight: 178,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23111092
CAS-Nummer: 77458-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR