3-Fluoro-4-(methylsulphonyl)phenol 95%

Lieferant: Apollo Scientific

PC910306-1G
APOSPC910306-1GEA 632 EUR
APOSPC910306-1G
3-Fluoro-4-(methylsulphonyl)phenol 95%
3-Fluoro-4-(methylsulphonyl)phenol
Formel: C₇H₇FO₃S
Molecular Weight: 190.192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20258998
CAS-Nummer: 859538-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR