2-Hydroxy-4-(trifluoromethoxy)benzaldehyde 98%

Lieferant: Apollo Scientific
Achtung

PC910312-1G PC910312-5G
APOSPC910312-1GEA 377 EUR
APOSPC910312-1G APOSPC910312-5G
2-Hydroxy-4-(trifluoromethoxy)benzaldehyde 98%
2-Hydroxy-4-(trifluoromethoxy)benzaldehyde
Formel: C₈H₅F₃O₃
Molecular Weight: 206.119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18397426
CAS-Nummer: 1071156-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR