4-Chloro-2-methyl-5-(trifluoromethyl)aniline 95%

Lieferant: Apollo Scientific
Achtung

PC910371-1G PC910371-250MG
APOSPC910371-1GEA 481 EUR
APOSPC910371-1G APOSPC910371-250MG
4-Chloro-2-methyl-5-(trifluoromethyl)aniline 95%
4-Chloro-2-methyl-5-(trifluoromethyl)aniline
Formel: C₈H₇ClF₃N
Lagertemperatur: Kühlschrank
MDL: MFCD09907679
CAS-Nummer: 1379346-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR