3-Fluor-5-iodbenzonitril 98%

Lieferant: Apollo Scientific

PC9104-1G
APOSPC9104-1GEA 79.9 EUR
APOSPC9104-1G
3-Fluor-5-iodbenzonitril 98%
3-Fluor-5-iodbenzonitril
Formel: C₇H₃FIN
Molecular Weight: 247,0100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116468
CAS-Nummer: 723294-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR