5-Fluoro-6-methyl-1,3-benzothiazol-2-amine 95%

Lieferant: Apollo Scientific

PC910401-1G
APOSPC910401-1GEA 632 EUR
APOSPC910401-1G
5-Fluoro-6-methyl-1,3-benzothiazol-2-amine 95%
5-Fluoro-6-methyl-1,3-benzothiazol-2-amine
Formel: C₈H₇FN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD08669847
CAS-Nummer: 1155287-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR