2-Fluoro-4-(methylamino)benzonitrile 95%

Lieferant: Apollo Scientific

PC910446-1G
APOSPC910446-1GEA 754 EUR
APOSPC910446-1G
2-Fluoro-4-(methylamino)benzonitrile 95%
2-Fluoro-4-(methylamino)benzonitrile
Formel: C₈H₇FN₂
Molecular Weight: 150,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18826265
CAS-Nummer: 171049-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR