Methyl 6-fluoro-1H-indazole-3-carboxylate 95%

Lieferant: Apollo Scientific

PC910615-1G
APOSPC910615-1GEA 36.4 EUR
APOSPC910615-1G
Methyl 6-fluoro-1H-indazole-3-carboxylate 95%
Methyl 6-fluoro-1H-indazole-3-carboxylate
Formel: C₉H₇FN₂O₂
Molecular Weight: 194.162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371604
CAS-Nummer: 885279-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR