3-(Chloromethyl)-2-fluoropyridine 98%

Lieferant: Apollo Scientific

PC910721-1G PC910721-5G
APOSPC910721-1GEA 0 EUR
APOSPC910721-1G APOSPC910721-5G
3-(Chloromethyl)-2-fluoropyridine 98%
3-(Chloromethyl)-2-fluoropyridine
Formel: C₆H₅ClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD10697617
CAS-Nummer: 315180-14-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR