3,6-Dibromo-2-fluoropyridine 98%

Lieferant: Apollo Scientific

PC910732-1G
APOSPC910732-1GEA 719 EUR
APOSPC910732-1G
3,6-Dibromo-2-fluoropyridine 98%
3,6-Dibromo-2-fluoropyridine
Formel: C₅H₂Br₂FN
Molecular Weight: 254,882 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18258101
CAS-Nummer: 1256820-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR