2-Chloro-6-fluoropyridin-4-ol 98%

Lieferant: Apollo Scientific

PC910734-1G
APOSPC910734-1GEA 661 EUR
APOSPC910734-1G
2-Chloro-6-fluoropyridin-4-ol 98%
2-Chloro-6-fluoropyridin-4-ol
Formel: C₅H₃ClFNO
Molecular Weight: 147,535 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD26383422
CAS-Nummer: 1807206-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR