6-Chloro-5-fluoropyridin-3-amine 98%

Lieferant: Apollo Scientific
Achtung

PC910736-250MG
APOSPC910736-250MGEA 0 EUR
APOSPC910736-250MG
6-Chloro-5-fluoropyridin-3-amine 98%
6-Chloro-5-fluoropyridin-3-amine
Formel: C₅H₄ClFN₂
Lagertemperatur: Kühlschrank
MDL: MFCD18257897
CAS-Nummer: 1256806-83-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR