(E)-N-[(5-fluoropyridin-3-yl)methylidene]hydroxylamine 97%

Lieferant: Apollo Scientific

PC910742-1G
APOSPC910742-1GEA 451 EUR
APOSPC910742-1G
(E)-N-[(5-fluoropyridin-3-yl)methylidene]hydroxylamine 97%
(E)-N-[(5-fluoropyridin-3-yl)methylidene]hydroxylamine
Formel: C₆H₅FN₂O
Molecular Weight: 140,115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26940391
CAS-Nummer: 1198353-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR