(2,6-Dichloro-5-fluoropyridin-3-yl)methanamine 96%

Lieferant: Apollo Scientific

PC910760-1G
APOSPC910760-1GEA 451 EUR
APOSPC910760-1G
(2,6-Dichloro-5-fluoropyridin-3-yl)methanamine 96%
(2,6-Dichloro-5-fluoropyridin-3-yl)methanamine
Formel: C₆H₅Cl₂FN₂
Molecular Weight: 195,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213012
CAS-Nummer: 771581-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR