2-Methoxy-6-(trifluoromethyl)pyridin-4-ol 98%
Lieferant: Apollo Scientific
PC910764-1G
APOSPC910764-1GEA
632
EUR
APOSPC910764-1G
2-Methoxy-6-(trifluoromethyl)pyridin-4-ol 98%
2-Methoxy-6-(trifluoromethyl)pyridin-4-ol
Formel:
C₇H₆F₃NO₂ Molecular Weight: 193,123 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16611389 CAS-Nummer: 1227513-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...