N-Ethyl-3-fluoroaniline 95%

Lieferant: Apollo Scientific

PC910847-1G PC910847-250MG
APOSPC910847-1GEA 0 EUR
APOSPC910847-1G APOSPC910847-250MG
N-Ethyl-3-fluoroaniline 95%
N-Ethyl-3-fluoroaniline
Formel: C₈H₁₀FN
Lagertemperatur: Raumtemperatur
MDL: MFCD11144550
CAS-Nummer: 2707-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR