2-fluorobenzene-1,3-dicarboxylic acid 95%

Lieferant: Apollo Scientific

PC910889-1G
APOSPC910889-1GEA 159 EUR
APOSPC910889-1G
2-fluorobenzene-1,3-dicarboxylic acid 95%
2-fluorobenzene-1,3-dicarboxylic acid
Formel: C₈H₅FO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00042505
CAS-Nummer: 1583-65-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR